Product Details

SNP ID
rs13481536
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:75453058 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ATATGAAGTCCAACATTCCCTTCGC[C/T]ACCATGCTTATCACCTCTTCTTTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Ppp2r5e PubMed Links

Gene Details

Gene
Ppp2r5e
Gene Name
protein phosphatase 2, regulatory subunit B', epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012024.2 2633 UTR 3 NP_036154.1

View Full Product Details