Product Details
- SNP ID
-
rs13476498
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:52245290 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCTGTCACTACTCACGTCACTGGC[A/G]ATATCTCTGGAGGCCTTGGCCGCCA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
Neb
PubMed Links
Gene Details
- Gene
- Neb
- Gene Name
- nebulin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_010889.1 |
10226 |
Silent Mutation |
|
|
NP_035019.1 |
View Full Product Details