Product Details
- SNP ID
-
rs13481953
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:95008925 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTACATGAATGTATACATACACGT[A/G]TACCAGGAAGACAGAAGCGGTTTTA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Wdr41
PubMed Links
Gene Details
- Gene
- Wdr41
- Gene Name
- WD repeat domain 41
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_172590.3 |
|
Intron |
|
|
NP_766178.2 |
View Full Product Details