Product Details
- SNP ID
-
rs13477559
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:11221235 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAACAACTTCCCAGAGGTCTTTAGC[A/G]GTCCGTCTACTCTCTTTAGGGCCTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Ints8
PubMed Links
Gene Details
- Gene
- Ints8
- Gene Name
- integrator complex subunit 8
View Full Product Details