Product Details

SNP ID
rs3699842
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.6:32837759 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GCCTCTCTCACCCAGCAGCTTTGCT[C/G]AGATCTATGCTGGTAGCAGGCTGGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
Chchd3 PubMed Links

Gene Details

Gene
Chchd3
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025336.1 Intron NP_079612.1

View Full Product Details