Product Details
- SNP ID
-
rs13476567
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:70989873 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGAAACACTTAGAAAGGGCTGTCA[C/T]AGCAATGGCAAAGCAGGCACCTCAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Mettl8
PubMed Links
Gene Details
- Gene
- Mettl8
- Gene Name
- methyltransferase like 8
View Full Product Details