Product Details

SNP ID
rs13480518
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:18159754 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTCAACAAGATACTTACAGAC[C/T]CTTCTTTTGACAGCTCCATCAGTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Ect2l PubMed Links

Gene Details

Gene
Ect2l
Gene Name
epithelial cell transforming sequence 2 oncogene-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195036.1 1762 Missense Mutation NP_001181965.1

View Full Product Details