Product Details

SNP ID
rs13481044
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:57831274 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCCTTTTCCGCTTGCTTTCGCGACC[A/G]CCATCCGTCTTTTTGAGTTCAGCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Hand1 PubMed Links

Gene Details

Gene
Hand1
Gene Name
heart and neural crest derivatives expressed transcript 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_008213.2 874 Silent Mutation NP_032239.1

View Full Product Details