Product Details

SNP ID
rs13478661
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.6:24148233 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTCAAATGTTTTATTTATAAATGTA[C/T]TTTCAACTATAGGGATAGTAATATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Slc13a1 PubMed Links

Gene Details

Gene
Slc13a1
Gene Name
solute carrier family 13 (sodium/sulfate symporters), member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019481.2 Intron NP_062354.2

View Full Product Details