Product Details

SNP ID
rs4174048
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.16:37536991 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGACCCCAGTCAGACGAGCTGTTTG[G/T]GTGCTGTAGGAGCCTGAAAAGTCCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Gtf2e1 PubMed Links

Gene Details

Gene
Gtf2e1
Gene Name
general transcription factor II E, polypeptide 1 (alpha subunit)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_028812.3 Intron NP_083088.1
Gene
Rabl3
Gene Name
RAB, member RAS oncogene family-like 3
There are no transcripts associated with this gene.

View Full Product Details