Product Details
- SNP ID
-
rs4179504
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:44259475 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTGATGACCACAGCGAAGGAGGC[A/G]TAAGCAGAGTAGGCGCTGGCGTTGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Sidt1
PubMed Links
Gene Details
- Gene
- Sidt1
- Gene Name
- SID1 transmembrane family, member 1
View Full Product Details