Product Details
- SNP ID
-
rs13477815
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:85092716 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGTGGAGTCTTCATCTGACATCA[C/T]GAGCTTGGCAGAGGAAGTTTCTAGG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
Cntln
PubMed Links
Gene Details
- Gene
- Cntln
- Gene Name
- centlein, centrosomal protein
View Full Product Details