Product Details

SNP ID
rs13459188
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:76352871 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GACTGAGCAAGAGCCAGGGTGGTGA[G/T]GATGAGAGTCCTCTGAGCGACAAGT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Cyc1 PubMed Links

Gene Details

Gene
Cyc1
Gene Name
cytochrome c-1
There are no transcripts associated with this gene.

Gene
Maf1
Gene Name
MAF1 homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164607.1 606 Missense Mutation NP_001158079.1
NM_001164608.1 606 Missense Mutation NP_001158080.1
NM_026859.3 606 Missense Mutation NP_081135.3
Gene
Sharpin
Gene Name
SHANK-associated RH domain interacting protein
There are no transcripts associated with this gene.

View Full Product Details