Product Details

SNP ID
rs13483336
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.18:45339928 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GTTATTATAAGTATATTAAGGTCTC[A/C]AAAACCATGATCTGTCCTCTGAGAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Kcnn2 PubMed Links

Gene Details

Gene
Kcnn2
Gene Name
potassium intermediate/small conductance calcium-activated channel, subfamily N, member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080465.2 Intron NP_536713.1

View Full Product Details