Product Details

SNP ID
rs13481240
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:113655729 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACCTGACCCTGTGTGGGCATTTCAC[A/G]GTCAGGCTTTAAATGGTCACTGTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Cog1 PubMed Links

Gene Details

Gene
Cog1
Gene Name
component of oligomeric golgi complex 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013581.3 Intron NP_038609.3
Gene
Fam104a
Gene Name
family with sequence similarity 104, member A
There are no transcripts associated with this gene.

Gene
Slc39a11
Gene Name
solute carrier family 39 (metal ion transporter), member 11
There are no transcripts associated with this gene.

View Full Product Details