Product Details
- SNP ID
-
rs13482944
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:30445981 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAGTATGGTTCACAACTGAGCCAA[A/C]AGTGAGATAACAACTCACATCTTGG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Btbd9
PubMed Links
Gene Details
- Gene
- Btbd9
- Gene Name
- BTB (POZ) domain containing 9
View Full Product Details