Product Details

SNP ID
rs13459054
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:165215752 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GCTGTACCACATTTTATTCTGAGGG[C/G]GTACTTTGTGAGTCCACTGGGTTAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
Tiprl PubMed Links

Gene Details

Gene
Tiprl
Gene Name
TIP41, TOR signalling pathway regulator-like (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145513.4 929 Silent Mutation NP_663488.1

View Full Product Details