Product Details

SNP ID
rs13480625
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:64710348 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GAGGAAAGGGGGAATCAGAAGAGAC[A/G]GAGACTAAACAGGATTGATGGACAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Ctnna3 PubMed Links

Gene Details

Gene
Ctnna3
Gene Name
catenin (cadherin associated protein), alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164376.1 Intron NP_001157848.1
NM_001164519.1 Intron NP_001157991.1
NM_177612.3 Intron NP_808280.2

View Full Product Details