Product Details
- SNP ID
-
rs13480943
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:31365493 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTGCTGATGCAGCCAAATTTATG[A/C]CGCAGGGCATGGGCCTTGCACCTCA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Stc2
PubMed Links
Gene Details
- Gene
- Stc2
- Gene Name
- stanniocalcin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_011491.3 |
544 |
Silent Mutation |
|
|
NP_035621.1 |
View Full Product Details