Product Details

SNP ID
rs6168834
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:150837612 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CATCCGGCCCCGTAGATCAGATGGG[A/G]CTTCCACAGCTCACGTGCAACCACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Abhd12 PubMed Links

Gene Details

Gene
Abhd12
Gene Name
abhydrolase domain containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024465.3 Intron NP_077785.2
Gene
Pygb
Gene Name
brain glycogen phosphorylase
There are no transcripts associated with this gene.

View Full Product Details