Product Details
- SNP ID
-
rs6194543
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:155741792 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAAATTAAATTAGCGAGAAAGGAG[G/T]GTGTCTTACGTTGCATCGGCACACC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Lhx4
PubMed Links
Gene Details
- Gene
- Lhx4
- Gene Name
- LIM homeobox protein 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_010712.2 |
|
Intron |
|
|
NP_034842.2 |
View Full Product Details