Product Details

SNP ID
rs4222678
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:149872527 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TACAGTTAAAGGGATTATTATTTGA[C/T]GATGACGTACATTTTGAATTAGTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Pla2g4a PubMed Links

Gene Details

Gene
Pla2g4a
Gene Name
phospholipase A2, group IVA (cytosolic, calcium-dependent)
There are no transcripts associated with this gene.

View Full Product Details