Product Details

SNP ID
rs13477165
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.3:66198643 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACTTCATCTCTGTTTTTATGCATAG[C/G]TCAGCTGCCTTCTAGCTCTGGAAAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
Veph1 PubMed Links

Gene Details

Gene
Veph1
Gene Name
ventricular zone expressed PH domain-containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145820.3 Intron NP_665819.2

View Full Product Details