Product Details
- SNP ID
-
rs13481249
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:116541583 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTTCTCCTGTAGCTTCTTGATATC[A/G]TCCAAAAACTTCTTCTCTCGGGTTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Sphk1
PubMed Links
Gene Details
- Gene
- Sphk1
- Gene Name
- sphingosine kinase 1
There are no transcripts associated with this gene.
- Gene
- Ube2o
- Gene Name
- ubiquitin-conjugating enzyme E2O
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173755.3 |
2590 |
Silent Mutation |
|
|
NP_776116.2 |
View Full Product Details