Product Details
- SNP ID
-
rs13476404
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:29500710 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACACAACCCAATGACACGCTCCTT[A/G]ACTCTCACAAGGAACAGGCGGCCCT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Med27
PubMed Links
Gene Details
- Gene
- Med27
- Gene Name
- mediator complex subunit 27
There are no transcripts associated with this gene.
View Full Product Details