Product Details

SNP ID
rs13459189
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:79725489 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CGTGCACCCGGAAGCGGTAGATACA[C/T]GTGTACTCAGGGTGGCCCCAGTTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Gtpbp1 PubMed Links

Gene Details

Gene
Gtpbp1
Gene Name
GTP binding protein 1
There are no transcripts associated with this gene.

Gene
Sun2
Gene Name
Sad1 and UNC84 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205345.1 2292 Silent Mutation NP_001192274.1
NM_001205346.1 2292 Silent Mutation NP_001192275.1
NM_194342.3 2292 Silent Mutation NP_919323.2

View Full Product Details