Product Details
- SNP ID
-
rs13477023
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:25890396 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCACTTCCTTTAGACAAACTATTG[A/C]TGTGCATCTCAGTTAGAAATGTGCT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Nlgn1
PubMed Links
Gene Details
- Gene
- Nlgn1
- Gene Name
- neuroligin 1
View Full Product Details