Product Details
- SNP ID
-
rs13476899
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:167238562 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAAAAGAACTCTCAGATTGTCAGC[A/C]AACAAGAATCAGCCCAAACCTTGCT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Ptgis
PubMed Links
Gene Details
- Gene
- Ptgis
- Gene Name
- prostaglandin I2 (prostacyclin) synthase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_008968.3 |
|
Intron |
|
|
NP_032994.1 |
View Full Product Details