Product Details
- SNP ID
-
rs13480792
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:120407822 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGAAATGCAGATTTTTCTGAATGA[A/G]TTCGGTTCCCTATTGCACGGTGTGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Hmga2
PubMed Links
Gene Details
- Gene
- Hmga2
- Gene Name
- high mobility group AT-hook 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_010441.2 |
|
Intron |
|
|
NP_034571.1 |
View Full Product Details