Product Details
- SNP ID
-
rs29527479
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:64762995 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAATTCTGGTATGATACATAAAGTG[A/G]AAAATTTTACACCTGATGTCTTGTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Cntnap3
PubMed Links
Gene Details
- Gene
- Cntnap3
- Gene Name
- contactin associated protein-like 3
View Full Product Details