Product Details
- SNP ID
-
rs13482156
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:45238727 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCAAATGTTTTTAATGAAGTGAT[A/G]ATTTATAAATTATTTTCAATGTTCT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
Gpr137c
PubMed Links
Gene Details
- Gene
- Gpr137c
- Gene Name
- G protein-coupled receptor 137C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_027518.2 |
|
Intron |
|
|
NP_081794.2 |
View Full Product Details