Product Details

SNP ID
rs13478721
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.6:39116753 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGGGGTAGGAGACAGGTTCAGGTAA[A/T]TGGGATATTTATCACTTTTAGACCT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
4930599N23Rik PubMed Links

Gene Details

Gene
4930599N23Rik
Gene Name
RIKEN cDNA 4930599N23 gene
There are no transcripts associated with this gene.

Gene
Parp12
Gene Name
poly (ADP-ribose) polymerase family, member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172893.3 Intron NP_766481.2

View Full Product Details