Product Details

SNP ID
rs13480737
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:103418322 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AAAGTGGTTCCCCAACACTGGATAC[C/T]GCTCCCAATGGACGCTATGGGATTG
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
Slc6a15 PubMed Links

Gene Details

Gene
Slc6a15
Gene Name
solute carrier family 6 (neurotransmitter transporter), member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252330.1 2490 Silent Mutation NP_001239259.1
NM_175328.3 2490 Silent Mutation NP_780537.1

View Full Product Details