Product Details

SNP ID
rs13482117
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.14:29478136 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGGGAACATCTGTCCCAGACAAGGT[G/T]AGCTGTGGTCATAGTAATAAGGTGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Cacna2d3 PubMed Links

Gene Details

Gene
Cacna2d3
Gene Name
calcium channel, voltage-dependent, alpha2/delta subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_009785.1 Intron NP_033915.1

View Full Product Details