Product Details

SNP ID
rs13480685
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:86123106 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGGTCTCCCTTGTATTTCAGGCCTC[A/T]CCCTGGAAGGGGTGTTGGTGGGATA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
4930486F22Rik PubMed Links

Gene Details

Gene
4930486F22Rik
Gene Name
RIKEN cDNA 4930486F22 gene
There are no transcripts associated with this gene.

Gene
Syn3
Gene Name
synapsin III
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164495.1 Intron NP_001157967.1
NM_013722.3 Intron NP_038750.2

View Full Product Details