Product Details

SNP ID
rs13482590
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:55113379 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGACATTCCCATCGCTTCCTGTTCC[C/T]TTTTCTTGGGCATATGCTTAAGACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Deptor PubMed Links

Gene Details

Gene
Deptor
Gene Name
DEP domain containing MTOR-interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037937.3 Intron NP_001033026.1
NM_145470.3 Intron NP_663445.2
Gene
Gm9920
Gene Name
predicted gene 9920
There are no transcripts associated with this gene.

View Full Product Details