Product Details

SNP ID
rs13481787
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:43544457 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ATACTTGGACGTGAAGCGCCTTACC[C/T]GCTAAGTATTTGATTGTATCAAGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Mcur1 PubMed Links

Gene Details

Gene
Mcur1
Gene Name
mitochondrial calcium uniporter regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081059.3 1031 Silent Mutation NP_001074528.1

View Full Product Details