Product Details
- SNP ID
-
rs13475705
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:5595513 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTTCTGATTGTGAGCTCGTCTAT[C/T]AGTTTGGCAGGTTTTTAAGCACTTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Oprk1
PubMed Links
Gene Details
- Gene
- Oprk1
- Gene Name
- opioid receptor, kappa 1
View Full Product Details