Product Details
- SNP ID
-
rs13481398
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:38632034 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACGATTTATGTTTTCAGACGCCAC[G/T]CCATCAGGAAAACGAGGGCTTGAAA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Dgkb
PubMed Links
Gene Details
- Gene
- Dgkb
- Gene Name
- diacylglycerol kinase, beta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178681.4 |
4168 |
UTR 3 |
|
|
NP_848796.2 |
View Full Product Details