Product Details
- SNP ID
-
rs6353593
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:68424814 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGTCCAGTATGGACCAAGCATTA[G/T]TTAAGCTGATGGGAAGGAAGCAGAG
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Stk39
PubMed Links
Gene Details
- Gene
- Stk39
- Gene Name
- serine/threonine kinase 39
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016866.2 |
|
Intron |
|
|
NP_058562.1 |
View Full Product Details