Product Details
- SNP ID
-
rs13476403
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:29245683 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAAGCACTCCCCCGTGCCCGTGCT[C/T]GCTAGTGCCTGGCCCAGCTGTCTCC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
6530402F18Rik
PubMed Links
Gene Details
- Gene
- 6530402F18Rik
- Gene Name
- RIKEN cDNA 6530402F18 gene
There are no transcripts associated with this gene.
- Gene
- Ntng2
- Gene Name
- netrin G2
There are no transcripts associated with this gene.
View Full Product Details