Product Details
- SNP ID
-
rs13483366
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:53743285 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTCTTATTCCAAACTCAGAAACTA[A/T]TTTTACACTGTGTTTTTTTTTTTTT
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Cep120
PubMed Links
Gene Details
- Gene
- Cep120
- Gene Name
- centrosomal protein 120
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178686.3 |
|
Intron |
|
|
NP_848801.2 |
View Full Product Details