Product Details

SNP ID
rs13477682
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.4:46540499 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGGACTTCTCTCCCAGATGATGCTC[C/T]GCGGTCCACTTTGCTAGCTTCTCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Coro2a PubMed Links

Gene Details

Gene
Coro2a
Gene Name
coronin, actin binding protein 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164804.1 1475 Silent Mutation NP_001158276.1
NM_178893.4 1475 Silent Mutation NP_849224.2
Gene
Gm16731
Gene Name
predicted gene, 16731
There are no transcripts associated with this gene.

Gene
Trim14
Gene Name
tripartite motif-containing 14
There are no transcripts associated with this gene.

View Full Product Details