Product Details

SNP ID
rs13478127
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.5:20250210 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GAAAAAAGACTTCCCTTTCCTTTCA[G/T]CCCTAGGGTAAAGGGCAAAACGTAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Magi2 PubMed Links

Gene Details

Gene
Magi2
Gene Name
membrane associated guanylate kinase, WW and PDZ domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170745.1 Intron NP_001164216.1
NM_001170746.1 Intron NP_001164217.1
NM_015823.3 Intron NP_056638.1

View Full Product Details