Product Details

SNP ID
rs13483739
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.X:48416800 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GACTTTTGGCTTCTCCCAGGGAGGG[C/T]TATCCTTGTCCTCGGGGGAGGCTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Bcorl1 PubMed Links

Gene Details

Gene
Bcorl1
Gene Name
BCL6 co-repressor-like 1
There are no transcripts associated with this gene.

Gene
Elf4
Gene Name
E74-like factor 4 (ets domain transcription factor)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019680.2 1296 Missense Mutation NP_062654.2

View Full Product Details