Product Details

SNP ID
rs33221062
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:44637733 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GTTCAGCTGTTTAAGGTTGTATACA[G/T]CGGTTTCCATGGTGAATCTATATAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Runx2 PubMed Links

Gene Details

Gene
Runx2
Gene Name
runt related transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145920.2 Intron NP_001139392.1
NM_001146038.2 Intron NP_001139510.1
NM_001271627.1 Intron NP_001258556.1
NM_001271630.1 Intron NP_001258559.1
NM_001271631.1 Intron NP_001258560.1
NM_009820.5 Intron NP_033950.2

View Full Product Details