Product Details

SNP ID
rs4223511
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:131993535 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCCACGTTACAAAGCTGCGTTTGTC[C/T]ATGGTGTTAGCTGTGTGCCCTTCAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Rassf2 PubMed Links

Gene Details

Gene
Rassf2
Gene Name
Ras association (RalGDS/AF-6) domain family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175445.4 4100 UTR 3 NP_780654.1

View Full Product Details