Product Details

SNP ID
rs4185176
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.16:49891852 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGGAAGAGTCGCTAATGAACGCTTG[A/T]TTGAATGGGGTAGGTTTGAAACAGC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Cd47 PubMed Links

Gene Details

Gene
Cd47
Gene Name
CD47 antigen (Rh-related antigen, integrin-associated signal transducer)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_010581.3 Intron NP_034711.1

View Full Product Details