Product Details

SNP ID
rs13483021
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:50733345 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCTCTTTTCAACAAGAGAGAAGCAG[C/G]TTTTGGGCCTAGCATGGAGGAGGCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
Tbc1d5 PubMed Links

Gene Details

Gene
Tbc1d5
Gene Name
TBC1 domain family, member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285991.1 5365 UTR 3 NP_001272920.1
NM_001285993.1 5365 UTR 3 NP_001272922.1
NM_028162.4 5365 UTR 3 NP_082438.3

View Full Product Details