Product Details

SNP ID
rs6163041
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:87702931 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CACTAGAACTAGACTCCATGATTTC[C/T]GCAACATTCTTACTTTGACTCAGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Fam19a5 PubMed Links

Gene Details

Gene
Fam19a5
Gene Name
family with sequence similarity 19, member A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252310.1 Intron NP_001239239.1
NM_134096.2 Intron NP_598857.1

View Full Product Details